accessibility menu, dialog, popup

UserName

CAS RN 6967-89-1

IUPAC Name: naphthalene-2-carbothioamide
Synonyms: naphthalene-2-carbothioamide
Molecular Weight (g/mol): 187.26
Molecular Formula: C11H9NS
InChi Key: ZQGJZFKITDDUEH-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results