accessibility menu, dialog, popup

UserName

CH3CMgCl

CAS RN 696-61-7

IUPAC Name: magnesium(2+) 4-methylbenzen-1-ide chloride
Synonyms: magnesium(2+) 4-methylbenzen-1-ide chloride
Molecular Weight (g/mol): 150.89
Molecular Formula: C7H7ClMg
InChi Key: BLMRDOZWELZHDX-UHFFFAOYSA-M
SMILES: [Mg++].[Cl-].CC1=CC=[C-]C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results