CAS RN 696-44-6
IUPAC Name:
N,3-dimethylaniline
Synonyms:
benzenamine, N,3-dimethyl-
Molecular Weight (g/mol):
121.18
Molecular Formula:
C8H11N
InChi Key:
FBGJJTQNZVNEQU-UHFFFAOYSA-N
SMILES:
CNC1=CC=CC(C)=C1
1
–
2
of
2
results
1
N,3-Dimethylaniline, TRC
CAS: 696-44-6 Molecular Formula: C8 H11 N Molecular Weight (g/mol): 121.18 Synonym: N,3-Dimethylaniline,Tolnaftate Imp. D (EP),N-Methyl-m-toluidine,Benzenamine, N,3-dimethyl-,m-Toluidine, N-methyl- (6CI,7CI,8CI),N,3-Dimethylbenzenamine,N-(3-Methylphenyl)methylamine,N-Methyl(3-methylphenyl)amine,N-Methyl-N-(3-methylphenyl)amine,N-Methyl-N-(m-tolyl)amine,NSC 9396,m,N-Dimethylaniline IUPAC Name: N,3-dimethylaniline SMILES: CNc1cccc(C)c1
2
N,3-Dimethylaniline (N-Methyl-m-toluidine), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.