CAS RN 6946-18-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
D-glycero-D-gulo-Heptopyranose, TRC
CAS: 6946-18-5 Molecular Formula: C7H14O7 Molecular Weight (g/mol): 210.18 Synonym: NSC 57733 IUPAC Name: (3R,4R,5R,6R)-6-[(1R)-1,2-dihydroxyethyl]oxane-2,3,4,5-tetrol SMILES: OC[C@@H](O)[C@H]1OC(O)[C@H](O)[C@H](O)[C@H]1O