CAS RN 694-48-4
IUPAC Name:
1,3-dimethyl-1H-pyrazole
Synonyms:
1,3-dimethylpyrazole
Molecular Weight (g/mol):
96.13
Molecular Formula:
C5H8N2
InChi Key:
NODLZCJDRXTSJO-UHFFFAOYSA-N
SMILES:
CN1C=CC(C)=N1
1
–
1
of
1
results
1
1,3-Dimethylpyrazole, TRC
CAS: 694-48-4 Molecular Formula: C5H8N2 Molecular Weight (g/mol): 96.13 Synonym: 2,5-Dimethylpyrazole,NSC 190569 IUPAC Name: 1,3-dimethylpyrazole SMILES: Cc1ccn(C)n1