CAS RN 693793-87-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Difluoro Atorvastatin Acetonide tert-Butyl Ester, TRC
CAS: 693793-87-2 Molecular Formula: C40H46F2N2O5 Molecular Weight (g/mol): 672.8 Synonym: (4R,6R)-6-[2-[2,3-Bis(4-fluorophenyl)-5-(1-methylethyl)-4-[(phenylamino)carbonyl]-1H-pyrrol-1-yl]ethyl]-2,2-dimethyl-1,3-Dioxane-4-acetic Acid 1,1-Dimethylethyl Ester IUPAC Name: tert-butyl 2-((4R,6R)-6-(2-(2,3-bis(4-fluorophenyl)-5-isopropyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)ethyl)-2,2-dimethyl-1,3-dioxan-4-yl)acetate SMILES: O=C(NC1=CC=CC=C1)C2=C(C(C)C)N(CC[C@@H](OC(C)(C)O3)C[C@@H]3CC(OC(C)(C)C)=O)C(C4=CC=C(F)C=C4)=C2C5=CC=C(F)C=C5