accessibility menu, dialog, popup

UserName

H3CONNNH2

CAS RN 6905-47-1

IUPAC Name: 6-methoxypyrazin-2-amine
Synonyms: pyrazinamine, 6-methoxy-
Molecular Weight (g/mol): 125.13
Molecular Formula: C5H7N3O
InChi Key: YGFNVCMGUHJLRU-UHFFFAOYSA-N
SMILES: COC1=NC(N)=CN=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results