accessibility menu, dialog, popup

UserName

CAS RN 68310-13-4

IUPAC Name: potassium palladium sulfite
Synonyms: potassium palladium sulfite
Molecular Weight (g/mol): 225.58
Molecular Formula: KO3PdS
InChi Key: FULLCBDUCZPCNB-UHFFFAOYSA-L
SMILES: [K+].[Pd].[O-]S([O-])=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results