CAS RN 67944-25-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Prazosin Hydrochloride Dihydrate, TRC
CAS: 67944-25-6 Molecular Formula: C19H21N5O4 • HCl • 2(H2O) Molecular Weight (g/mol): 455.9 Synonym: [4-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-1-piperazinyl]-2-furanylmethanone Hydrochloride Dihydrate,1-(4-Amino-6,7-dimethoxy-2-quinazolinyl)-4-(2-furanylcarbonxyl) IUPAC Name: (4-(4-amino-6,7-dimethoxyquinazolin-2-yl)piperazin-1-yl)(furan-2-yl)methanone hydrochloride dihydrate SMILES: O=C(N1CCN(CC1)C2=NC3=C(C=C(C(OC)=C3)OC)C(N)=N2)C4=CC=CO4.Cl.O.O