CAS RN 67372-68-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6,7-Demethylene Drospirenone, TRC
CAS: 67372-68-3 Molecular Formula: C23 H30 O3 Molecular Weight (g/mol): 354.48 Synonym: (15α,16α,17α)-15,16-dihydro-17-hydroxy-3-oxo-3'H-cyclopropa[15,16]pregna-4,15-diene-21-carboxylic acid γ-lactone,15β,16β-Methylene-3-oxo-17α-pregn-4-ene-21,17-carbolactone,17β-Hydroxy-15β,16β-methylene-3-oxo-17α-pregn-4-ene-21-carboxylic acid γ-lactone,3-oxo-15α,16α-dihydro-3′H-cyclopropa[15,16]-17α-pregn-4-ene-21,17-carbolactone (6,7-desmethylenedrospirenone),Drospirenone Imp. A (EP) SMILES: O=C1CC[C@]2(C)[C@@]3([H])CC[C@]4(C)[C@@]5(OC(CC5)=O)[C@@H](C6)[C@@H]6[C@@]4([H])[C@]3([H])CCC2=C1