accessibility menu, dialog, popup

UserName

CAS RN 67344-77-8

IUPAC Name: [(3-bromophenyl)methyl](methyl)amine
Synonyms: [(3-bromophenyl)methyl](methyl)amine
Molecular Weight (g/mol): 200.08
Molecular Formula: C8H10BrN
InChi Key: QCEANFBGRBLXHN-UHFFFAOYSA-N
SMILES: CNCC1=CC=CC(Br)=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results