CAS RN 66753-06-8
CAS RN 66753-06-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Amino-6-[(1,1-Dimethylethyl)Amino]-1,3,5-Triazin-2(1H)-One, TRC
CAS: 66753-06-8 Molecular Formula: C7 H13 N5 O Molecular Weight (g/mol): 183.21 Synonym: Desethylhydroxyterbuthylazine,Terbuthylazine metabolite IUPAC Name: 4-amino-6-(tert-butylamino)-1,3,5-triazin-2-ol SMILES: CC(C)(C)Nc1nc(N)nc(O)n1