accessibility menu, dialog, popup

UserName

CAS RN 6631-37-4

HNN

CAS RN 6631-37-4

IUPAC Name: N-phenylpyridin-2-amine
Synonyms: 2-pyridineamine, N-phenyl
Molecular Weight (g/mol): 170.22
Molecular Formula: C11H10N2
InChi Key: HUDSSSKDWYXKGP-UHFFFAOYSA-N
SMILES: N(C1=CC=CC=C1)C1=CC=CC=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results