CAS RN 66211-46-9
IUPAC Name:
3-aminopropane-1,2-diol
Synonyms:
3-aminopropane-1,2-diol
Molecular Weight (g/mol):
91.11
Molecular Formula:
C3H9NO2
InChi Key:
KQIGMPWTAHJUMN-UHFFFAOYNA-N
SMILES:
NCC(O)CO
1
–
1
of
1
results
1
(R)-3-Amino-1,2-propanediol, 98%
CAS: 66211-46-9 Molecular Formula: C3H9NO2 Molecular Weight (g/mol): 91.11 InChI Key: KQIGMPWTAHJUMN-GSVOUGTGSA-N Synonym: r-3-amino-1,2-propanediol,r-3-aminopropane-1,2-diol,r-+-3-amino-1,2-propandiol,2r-3-aminopropane-1,2-diol,r-+-3-amino-1,2-propanediol,r-+-amino-1,2-propanediol,1-aminoglycerol,1,2-propanediol, 3-amino-, 2r,pubchem5711,r-3-amino-1,2-propanol PubChem CID: 6994409 IUPAC Name: (2R)-3-aminopropane-1,2-diol SMILES: C(C(CO)O)N