CAS RN 66067-43-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
3-Ethylbenzophenone, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
3-Ethylbenzophenone, TRC
CAS: 66067-43-4 Molecular Formula: C15 H14 O Molecular Weight (g/mol): 210.27 Synonym: 3-Ethylbenzophenone,(3-Ethylphenyl)phenylmethanone,m-Ethylbenzophenone IUPAC Name: (3-ethylphenyl)-phenylmethanone SMILES: CCc1cccc(c1)C(=O)c2ccccc2