CAS RN 66003-55-2
CAS RN 66003-55-2
IUPAC Name:
sodium {1-[3-(methoxycarbonyl)-4,4-dimethyl-2,6-dioxocyclohexylidene]butyl}(prop-2-en-1-yloxy)azanide
Synonyms:
sodium {1-[3-(methoxycarbonyl)-4,4-dimethyl-2,6-dioxocyclohexylidene]butyl}(prop-2-en-1-yloxy)azanide
Molecular Weight (g/mol):
345.37
Molecular Formula:
C17H24NNaO5
InChi Key:
UUTYCSXRRNBAOR-UHFFFAOYNA-M
SMILES:
[Na+].CCCC([N-]OCC=C)=C1C(=O)CC(C)(C)C(C(=O)OC)C1=O