accessibility menu, dialog, popup

UserName

CAS RN 65273-47-4

IUPAC Name: [4-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)butyl]triphenylphosphanium bromide
Synonyms: [4-(1,3-dioxoisoindol-2-yl)butyl]triphenylphosphanium bromide
Molecular Weight (g/mol): 544.43
Molecular Formula: C30H27BrNO2P
InChi Key: YKNPTFHDEQEFAS-UHFFFAOYSA-M
SMILES: [Br-].O=C1N(CCCC[P+](C2=CC=CC=C2)(C2=CC=CC=C2)C2=CC=CC=C2)C(=O)C2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results