CAS RN 65109-70-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
S-Ethyl Ester-, (3-Hydroxypropyl)propyl- Carbamothioic Acid, TRC
CAS: 65109-70-8 Molecular Formula: C9 H19 N O2 S Molecular Weight (g/mol): 205.32 Synonym: Carbamothioic acid, (3-hydroxypropyl)propyl-, S-ethyl ester IUPAC Name: S-ethyl N-(3-hydroxypropyl)-N-propylcarbamothioate SMILES: CCCN(CCCO)C(=O)SCC