CAS RN 65101-87-3
IUPAC Name:
sodium 8-(9-hydroxy-2-{2-[6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-9-[(5-methoxy-6-methyloxan-2-yl)oxy]-4,10-dimethyl-1,6-dioxaspiro[4.5]decan-7-yl}-2,8-dimethyl-1,6-dioxaspiro[4.5]decan-7-yl)-2,4,6-trimethyl-5-oxonon-6-enoate
Synonyms:
sodium 8-(9-hydroxy-2-{2-[6-hydroxy-6-(hydroxymethyl)-3,5-dimethyloxan-2-yl]-9-[(5-methoxy-6-methyloxan-2-yl)oxy]-4,10-dimethyl-1,6-dioxaspiro[4.5]decan-7-yl}-2,8-dimethyl-1,6-dioxaspiro[4.5]decan-7-yl)-2,4,6-trimethyl-5-oxonon-6-enoate
Molecular Weight (g/mol):
889.11
Molecular Formula:
C47H77NaO14
InChi Key:
XMAIRYYXDCNFKP-UHFFFAOYNA-M
SMILES:
[Na+].COC1CCC(OC2CC(OC3(OC(CC3C)C3OC(O)(CO)C(C)CC3C)C2C)C2(C)CCC3(CC(O)C(C)C(O3)C(C)C=C(C)C(=O)C(C)CC(C)C([O-])=O)O2)OC1C
1
–
1
of
1
results
1
Nanchangmycin, MedChemExpress
MedChemExpress Nanchangmycin, a polyether antibiotic produced by Streptomyces nanchangensis NS3226, inhibits gram-positive bacteria. Nanchangmycin is a broad spectrum antiviral active against Zika virus.