accessibility menu, dialog, popup

UserName

NH2O(R)NH3

CAS RN 6485-67-2

IUPAC Name: (R)-carbamoyl(phenyl)methanaminium
Synonyms: (R)-carbamoyl(phenyl)methanaminium
Molecular Weight (g/mol): 151.19
Molecular Formula: C8H11N2O
InChi Key: KIYRSYYOVDHSPG-SSDOTTSWSA-O
SMILES: NC(=O)[C@H]([NH3+])C1=CC=CC=C1
Quantity 
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)