CAS RN 64742-88-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Turpentine Substitute, Extra Pure, SLR, Fisher Chemical™
CAS: 64742-88-7 MDL Number: 132767