accessibility menu, dialog, popup

UserName

CAS RN 644-35-9

CH3HO

CAS RN 644-35-9

IUPAC Name: 2-propylphenol
Synonyms: O-propylphenol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: LCHYEKKJCUJAKN-UHFFFAOYSA-N
SMILES: CCCC1=CC=CC=C1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results