CAS RN 642-36-4
CAS RN 642-36-4
IUPAC Name:
4,5-diphenyl-2,3-dihydro-1H-imidazol-2-one
Synonyms:
4,5-diphenyl-1,3-dihydroimidazol-2-one
Molecular Weight (g/mol):
236.27
Molecular Formula:
C15H12N2O
InChi Key:
JAWYWGRTWUVTEN-UHFFFAOYSA-N
SMILES:
O=C1NC(=C(N1)C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
4,5-Diphenylimidazolin-2-one, 98%, Thermo Scientific™
CAS: 642-36-4 Molecular Formula: C15H12N2O MDL Number: MFCD00022393 Synonym: 4,5-Diphenyl-2-glyoxalone