accessibility menu, dialog, popup

UserName

CAS RN 640296-19-1

IUPAC Name: 4-cyanopyridine-2-carboxylic acid
Synonyms: 4-cyanopyridine-2-carboxylic acid
Molecular Weight (g/mol): 148.12
Molecular Formula: C7H4N2O2
InChi Key: CMKZSIGRFONKKC-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results