CAS RN 6393-01-7
IUPAC Name:
2,5-dimethylbenzene-1,4-diamine
Synonyms:
4-amino-2,5-dimethylaniline
Molecular Weight (g/mol):
136.2
Molecular Formula:
C8H12N2
InChi Key:
BWAPJIHJXDYDPW-UHFFFAOYSA-N
SMILES:
CC1=CC(N)=C(C)C=C1N
1
–
1
of
1
results
1
2,5-Dimethyl-1,4-phenylenediamine, TRC
CAS: 6393-01-7 Molecular Formula: C8H12N2 Molecular Weight (g/mol): 136.2 SMILES: Cc1cc(N)c(C)cc1N