CAS RN 63740-81-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-7-Hydroxy Warfarin, TRC
CAS: 63740-81-8 Molecular Formula: C19 H16 O5 Molecular Weight (g/mol): 324.33 Synonym: 2H-1-Benzopyran-2-one, 4,7-dihydroxy-3-[(1S)-3-oxo-1-phenylbutyl]-,2H-1-Benzopyran-2-one, 4,7-dihydroxy-3-(3-oxo-1-phenylbutyl)-, (S)-,4,7-Dihydroxy-3-[(1S)-3-oxo-1-phenylbutyl]-2H-1-benzopyran-2-one,7-Hydroxy-S-warfarin,S-7-Hydroxywarfarin,(S)-7-Hydroxywarfarin IUPAC Name: 4,7-dihydroxy-3-[(1S)-3-oxo-1-phenylbutyl]chromen-2-one SMILES: CC(=O)C[C@H](C1=C(O)c2ccc(O)cc2OC1=O)c3ccccc3