accessibility menu, dialog, popup

UserName

CAS RN 63524-04-9

OOOOHOHOHH2ONa

CAS RN 63524-04-9

IUPAC Name: sodium 5-(1,2-dihydroxyethyl)-4-hydroxy-2-oxo-2,5-dihydrofuran-3-olate hydrate
Synonyms: sodium 5-(1,2-dihydroxyethyl)-4-hydroxy-2-oxo-5H-furan-3-olate hydrate
Molecular Weight (g/mol): 216.12
Molecular Formula: C6H9NaO7
InChi Key: NKESWEQTVZVSSF-UHFFFAOYNA-M
SMILES: O.[Na+].OCC(O)C1OC(=O)C([O-])=C1O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results