accessibility menu, dialog, popup

UserName

CAS RN 6336-82-9

IUPAC Name: 4-(naphthalen-2-yl)phenol
Synonyms: 4-(naphthalen-2-yl)phenol
Molecular Weight (g/mol): 220.27
Molecular Formula: C16H12O
InChi Key: NIRHUNSXEDESLN-UHFFFAOYSA-N
SMILES: OC1=CC=C(C=C1)C1=CC=C2C=CC=CC2=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results