accessibility menu, dialog, popup

UserName

H3CNN

CAS RN 6303-75-9

IUPAC Name: 2-pentylpyrazine
Synonyms: 2-pentylpyrazine
Molecular Weight (g/mol): 150.23
Molecular Formula: C9H14N2
InChi Key: KNDDHUQSPNJCKY-UHFFFAOYSA-N
SMILES: CCCCCC1=CN=CC=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results