accessibility menu, dialog, popup

UserName

ClClO

CAS RN 63024-77-1

IUPAC Name: 3-(chloromethyl)benzoyl chloride
Synonyms: 3-(chloromethyl)benzoyl chloride
Molecular Weight (g/mol): 189.04
Molecular Formula: C8H6Cl2O
InChi Key: YCAIYRWHKSJKEB-UHFFFAOYSA-N
SMILES: ClCC1=CC=CC(=C1)C(Cl)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)