accessibility menu, dialog, popup

UserName

H3CONNOOBr

CAS RN 62368-07-4

IUPAC Name: 1-(5-bromo-7-nitro-2,3-dihydro-1H-indol-1-yl)ethan-1-one
Synonyms: 1-(5-bromo-7-nitro-2,3-dihydroindol-1-yl)ethanone
Molecular Weight (g/mol): 285.1
Molecular Formula: C10H9BrN2O3
InChi Key: RCELVCGNAKOBPO-UHFFFAOYSA-N
SMILES: CC(=O)N1CCC2=CC(Br)=CC(=C12)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results