accessibility menu, dialog, popup

UserName

CH3NOO

CAS RN 62254-74-4

IUPAC Name: 5-methyl-1,2-oxazole-3-carbaldehyde
Synonyms: 5-methyl-1,2-oxazole-3-carbaldehyde
Molecular Weight (g/mol): 111.1
Molecular Formula: C5H5NO2
InChi Key: OCOCVXUEFWNRJU-UHFFFAOYSA-N
SMILES: CC1=CC(C=O)=NO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results