accessibility menu, dialog, popup

UserName

CH3FFFOHO

CAS RN 62089-35-4

IUPAC Name: 2-methyl-3-(trifluoromethyl)benzoic acid
Synonyms: 2-methyl-3-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: SSCFPSMXIDAJDT-UHFFFAOYSA-N
SMILES: CC1=C(C=CC=C1C(F)(F)F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results