CAS RN 617-92-5
IUPAC Name:
1-ethyl-1H-pyrrole
Synonyms:
1H-pyrrole, 1-ethyl-
Molecular Weight (g/mol):
95.15
Molecular Formula:
C6H9N
InChi Key:
VPUAYOJTHRDUTK-UHFFFAOYSA-N
SMILES:
CCN1C=CC=C1
1
–
1
of
1
results
1
1-Ethylpyrrole, TRC
CAS: 617-92-5 Molecular Formula: C6H9N Molecular Weight (g/mol): 95.14 Synonym: N-Ethylpyrrole IUPAC Name: 1-ethylpyrrole SMILES: CCn1cccc1