CAS RN 61612-83-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
delta8,9-Dehydro Estrone 3-Sulfate Sodium (Stabilized with up to 40% Tromethamine), TRC
CAS: 61612-83-7 Molecular Formula: C18 H19 O5 S . Na Molecular Weight (g/mol): 370.4 Synonym: Estra-1,3,5(10),8-tetraen-17-one, 3-(sulfooxy)-, sodium salt (1:1),Estra-1,3,5(10),8-tetraen-17-one, 3-(sulfooxy)-, sodium salt (9CI),δ8,9-Dehydroestrone sodium sulfate IUPAC Name: sodium;[(13S,14S)-13-methyl-17-oxo-7,11,12,14,15,16-hexahydro-6H-cyclopenta[a]phenanthren-3-yl] sulfate SMILES: [Na+].C[C@]12CCC3=C(CCc4cc(OS(=O)(=O)[O-])ccc34)[C@@H]1CCC2=O