CAS RN 61607-68-9
IUPAC Name:
1-[2-(dimethylamino)ethyl]-2,5-dihydro-1H-1,2,3,4-tetrazole-5-thione
Synonyms:
1-[2-(dimethylamino)ethyl]-2H-1,2,3,4-tetrazole-5-thione
Molecular Weight (g/mol):
173.24
Molecular Formula:
C5H11N5S
InChi Key:
ODDAWJGQWOGBCX-UHFFFAOYSA-N
SMILES:
CN(C)CCN1NN=NC1=S
1
–
1
of
1
results
1
1-(2-Dimethylaminoethyl)-5-mercaptotetrazole, TRC
CAS: 61607-68-9 Molecular Formula: C5H11N5S Molecular Weight (g/mol): 173.24 Synonym: Cefotiam Impurity 17 SMILES: CN(C)CCn1nnnc1S