CAS RN 615-21-4
IUPAC Name:
2-hydrazinyl-1,3-benzothiazole
Synonyms:
2-hydrazinobenzothiazole
Molecular Weight (g/mol):
165.21
Molecular Formula:
C7H7N3S
InChi Key:
JYSUYJCLUODSLN-UHFFFAOYSA-N
SMILES:
NNC1=NC2=CC=CC=C2S1
1
–
1
of
1
results
1
2-Hydrazinobenzothiazole, 97%
CAS: 615-21-4 Molecular Formula: C7H7N3S Molecular Weight (g/mol): 165.214 MDL Number: MFCD00041849 InChI Key: JYSUYJCLUODSLN-UHFFFAOYSA-N Synonym: 2-hydrazinobenzothiazole,2-hydrazinylbenzo d thiazole,2-hydrazino-1,3-benzothiazole,benzothiazolohydrazine,2 3h-benzothiazolone, hydrazone,benzothiazol-2-ylhydrazine,benzothiazole, 2-hydrazino,2-benzothiazolylhydrazine,usaf ek-3967,benzothiazol-2-yl-hydrazine PubChem CID: 11988 IUPAC Name: 1,3-benzothiazol-2-ylhydrazine SMILES: C1=CC=C2C(=C1)N=C(S2)NN