CAS RN 60919-46-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
15Alpha-Hydroxy-13Beta-ethyl-4-gonene-3,17-dione, TRC
CAS: 60919-46-2 Molecular Formula: C19H26O3 Molecular Weight (g/mol): 302.41 Synonym: 15alpha-Hydroxyl-13-ethyl-gon-4-en-3,17-dione,(15alpha)-13-Ethyl-15-hydroxy-gon-4-ene-3,17-dione,15alpha-hydroxy-18-methyl-estra-4-ene-3,17-dione IUPAC Name: (8R,9S,10R,13S,14S,15S)-13-ethyl-15-hydroxy-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthrene-3,17-dione SMILES: CC[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1[C@@H](O)CC2=O