CAS RN 60573-88-8
CAS RN 60573-88-8
IUPAC Name:
2-amino-2-(3-oxo-2,3-dihydro-1,2-oxazol-5-yl)acetic acid hydrate
Synonyms:
ibotenic acid hydrate
Molecular Weight (g/mol):
176.13
Molecular Formula:
C5H8N2O5
InChi Key:
QVKQQLYSTODTJN-UHFFFAOYNA-N
SMILES:
O.NC(C(O)=O)C1=CC(=O)NO1
No Refinements
1
–
0
of
0
results
1
(±)-Ibotenic acid monohydrate, 95%, Thermo Scientific Chemicals
CAS: 60573-88-8 Molecular Formula: C5H6N2O4·H2O Molecular Weight (g/mol): 176.1 MDL Number: MFCD00149579 InChI Key: QVKQQLYSTODTJN-UHFFFAOYSA-N Synonym: ibotenic acid hydrate,ibotenic acid monohydrate,pramuscimol,ibotensaeure,isotenic acid,ibotensaeure german,amino-3-hydroxy-5-isoxazolyl acetic acid,alpha-amino-3-hydroxy-5-isoxazolessigsaure hydrat,alpha-amino-3-hydroxy-5-isoxazoleacetic acid hydrate,alpha-amino-3-hydroxy-5-isoxazolessigsaure hydrat german PubChem CID: 43362 IUPAC Name: 2-amino-2-(3-oxo-1,2-oxazol-5-yl)acetic acid;hydrate SMILES: C1=C(ONC1=O)C(C(=O)O)N.O