accessibility menu, dialog, popup

UserName

CAS RN 6028-89-3

H3COS(E/Z)

CAS RN 6028-89-3

IUPAC Name: 1-(4-methylphenyl)-3-(thiophen-2-yl)prop-2-en-1-one
Synonyms: 1-(4-methylphenyl)-3-(thiophen-2-yl)prop-2-en-1-one
Molecular Weight (g/mol): 228.31
Molecular Formula: C14H12OS
InChi Key: OUDDTWPBSXBJOY-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C(=O)C=CC1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results