CAS RN 5959-86-4
CAS RN 5959-86-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(S)-1’-Methylnicotinium Iodide, TRC
CAS: 5959-86-4 Molecular Formula: C11H17N2I Molecular Weight (g/mol): 304.17 IUPAC Name: 3-[(2S)-1,1-dimethylpyrrolidin-1-ium-2-yl]pyridine;iodide SMILES: [I-].C[N+]1(C)CCC[C@H]1c2cccnc2