accessibility menu, dialog, popup

UserName

CAS RN 5905-69-1

FFOBr

CAS RN 5905-69-1

IUPAC Name: 1-bromo-4-(difluoromethoxy)benzene
Synonyms: 1-bromo-4-(difluoromethoxy)benzene
Molecular Weight (g/mol): 223.02
Molecular Formula: C7H5BrF2O
InChi Key: ORIYZUFTROJBQJ-UHFFFAOYSA-N
SMILES: FC(F)OC1=CC=C(Br)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results