CAS RN 58628-40-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bis(isopropylcyclopentadienyl)zirconium dichloride, 98%, Thermo Scientific™
CAS: 58628-40-3 Molecular Formula: C16H22Cl2Zr MDL Number: MFCD00269855