CAS RN 58176-73-1
IUPAC Name:
dithulium(3+) hydrate trioxalate
Synonyms:
dithulium(3+) hydrate trioxalate
Molecular Weight (g/mol):
619.94
Molecular Formula:
C6H2O13Tm2
InChi Key:
DUELVSSHRTZSBY-UHFFFAOYSA-H
SMILES:
O.[Tm+3].[Tm+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
1
–
1
of
1
results
1
Thulium(III) oxalate hydrate, REacton™, 99.9% (REO), Thermo Scientific™
CAS: 58176-73-1 Molecular Formula: C6O12Tm2 Molecular Weight (g/mol): 601.92 MDL Number: MFCD00050804 InChI Key: BSCZSHAJTJRUPP-UHFFFAOYSA-H Synonym: thulium oxalate hydrate,thulium iii oxalate hydrate,acmc-20ajie,ethanedioic acid,thulium 3+ salt,hydrate 3:2:? PubChem CID: 45052048 SMILES: [Tm+3].[Tm+3].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O