accessibility menu, dialog, popup

UserName

H3COOFFF

CAS RN 577-62-8

IUPAC Name: ethyl 2-(trifluoromethyl)benzoate
Synonyms: ethyl 2-(trifluoromethyl)benzoate
Molecular Weight (g/mol): 218.18
Molecular Formula: C10H9F3O2
InChi Key: BQLMZZVRHPZRBQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1C(F)(F)F
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results