CAS RN 57441-74-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6,7-Dimethoxy-1-indanone, TRC
CAS: 57441-74-4 Molecular Formula: C11H12O3 Molecular Weight (g/mol): 192.21 Synonym: 2,3-Dihydro-6,7-dimethoxy-1H-inden-1-one IUPAC Name: 6,7-dimethoxy-2,3-dihydro-1H-inden-1-one SMILES: O=C1CCC2=CC=C(OC)C(OC)=C21