CAS RN 574-12-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Isoflavone, TRC
CAS: 574-12-9 Molecular Formula: C15H10O2 Molecular Weight (g/mol): 222.24 Synonym: 3-Phenyl-4H-1-benzopyran-4-one,3-Phenyl-4-chromone,3-Phenylchromone,NSC 135405,Isoflavone 80 SMILES: O=C1C(=COc2ccccc12)c3ccccc3