CAS RN 573951-39-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4Beta,25-Dihydroxy Vitamin D3, TRC
CAS: 573951-39-0 Molecular Formula: C27H44O3 Molecular Weight (g/mol): 416.64 Synonym: (3beta,4beta,5E,7E)-9,10-Secocholesta-5,7,10(19)-triene-3,4,25-triol SMILES: C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2\C(=C\C=C/3\[C@@H](O)[C@@H](O)CCC3=C)\CCC[C@]12C