accessibility menu, dialog, popup

UserName

CAS RN 5702-74-9

H3CCH3NHCH3O

CAS RN 5702-74-9

IUPAC Name: N-[4-(propan-2-yl)phenyl]acetamide
Synonyms: N-(4-isopropylphenyl)acetamide
Molecular Weight (g/mol): 177.25
Molecular Formula: C11H15NO
InChi Key: BQZWLSZGPBNHDA-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(NC(C)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results