CAS RN 56824-22-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-(N-Methyl-2-pyrrolidinyl)ethyl Chloride Hydrochloride, TRC
CAS: 56824-22-7 Molecular Formula: C7 H14 Cl N . HCl Molecular Weight (g/mol): 184.1 Synonym: 2-(2-Chloroethyl)-1-methyl-pyrrolidine Hydrochloride,1-Methyl-2-(2-chloroethyl)pyrrolidine Hydrochloride,2-(2-Chloroethyl)-1-methylpyrrolidine Hydrochloride,2-(2-Chloroethyl)-N-methylpyrrolidine Hydrochloride,N-Methyl-2-(2-chloroethyl)pyrrolidine Hydrochloride IUPAC Name: 2-(2-chloroethyl)-1-methylpyrrolidine hydrochloride SMILES: ClCCC1N(CCC1)C.Cl